Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Lieferant: WILMAD LAB GLASS LUREX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
LG-6655-100
WILMLG-6655-100EA 784 EUR InStock
WILMLG-6655-100
Volatile oil apparatus for oils lighter than water, SP Wilmad-LabGlass
Destillationsbauteile Destillations-Bausätze
This distilling apparatus is used to determine volatile oil in drugs by distillation of a weighed quantity of drug in contact with water.
  • The Clevenger type receiver trap included in this apparatus is for use when the oils are lighter than water
  • The condenser rests on top of the receiver
  • There is a discharge stopcock below the return tube
  • Flask and receiver are connected by a standard taper 24/40 joint
The receiver, graduated to 5 ml with 0,1 ml increments, has a tube connecting the lower part below the scale with the top of the vapour tube to return excess water to the flask.
Lieferumfang: The apparatus also includes a 300 ml round bottom flask and a finger type condenser.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge