NMR Capillary tubes, SP Wilmad-LabGlass
Lieferant: WILMAD LAB GLASS LUREX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WG-1364-2-203M WG-1364-1.7 WG-1364-1 WG-1364-1.7-5 WG-1364-2 WG-1364-2.5A WG-1365-1
WILMWG-1364-2-203MEA 29.3 EUR InStock
WILMWG-1364-2-203M WILMWG-1364-1.7 WILMWG-1364-1 WILMWG-1364-1.7-5 WILMWG-1364-2 WILMWG-1364-2.5A WILMWG-1365-1
NMR Capillary tubes, SP Wilmad-LabGlass
Röhrchen NMR-Röhrchen
Pyrex® glass, Class A, ASTM type 1.
  • Tested at 400 MHz field
  • Ideal for small volume NMR measurement
  • With either both ends open or sealed at one end
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge