NMR tube carrier, SP Wilmad-LabGlass
Lieferant: WILMAD LAB GLASS LUREX
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
WG-6192
WILMWG-6192EA 51.3 EUR InStock
WILMWG-6192
NMR tube carrier, SP Wilmad-LabGlass
Transportbehälter Transportbehälter
Made from unbreakable polymer material with a snap-on cap, these tube carriers feature a shirt clip that keeps it secured in lab coat pockets.
  • Protection is provided for the sample and lab personnel during transport
  • Comes in a pack of three white NMR tube carriers
  • Holds 5×177,8 mm NMR tubes
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge