Pouring rings for laboratory bottles
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0.175.201
WITG0.175.201EA 0.4 EUR InStock
WITG0.175.201
Pouring rings for laboratory bottles
Verschlüsse Flaschenverschlüsse
Pouring rings, made of polypropylene.
  • For DIN GL threads
  • Heat resistant up to +140 °C
  • Good chemical resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge