Column head with valve stopcock acc. to Antlinger
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
1.610.029
WITG1.610.029EA 701 EUR InStock
WITG1.610.029
Column head with valve stopcock acc. to Antlinger
Destillationsbauteile Destillieraufsätze
Borosilicate glass3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Columns heads acc. to Antlinger, with PTFE-fine-dosing valve, cone to cloumn NS 29/32.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge