Chromatography column with disc
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2.133.009
WITG2.133.009EA 96.3 EUR InStock
WITG2.133.009
Chromatography column with disc
Chromatographiesäulen
Chromatography column made of borosilicate glass with fused in sintered glass disc with P0 porosity.
  • ST-PTFE-stopcock
  • Screw thread retaining nut
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge