Soxhlet extractors
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2.212.002 2.212.500
WITG2.212.002EA 446 EUR InStock
WITG2.212.002 WITG2.212.500
Soxhlet extractors
Extraktoren Extraktionsapparate, nach Soxhlet
Borosilicate glass. For extraction from solids with solvents.
  • Very good chemical resistance
  • Very good temperature resistance
The solvent is recirculated continuously and distilled from the extracted sample before contacting the solid again. The extract is concentrated in the distillation flask. The method allows an effective separation with relatively small amounts of extracting solvents.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge