Scheidetrichter, zylindrisch
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2.643.100
WITG2.643.100EA 55.9 EUR InStock
WITG2.643.100
Scheidetrichter, zylindrisch
Trichter Scheidetrichter
Cylindrical separatory funnels, made of borosilicat glass, with Witaflo PTFE needle valve stopcock.
  • Graduated
  • Very good chemical resistance
  • Very good temperature resistance
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge