NMR sample tube washer
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
2.900.029 2.900.032
WITG2.900.029EA 217 EUR InStock
WITG2.900.029 WITG2.900.032
NMR sample tube washer
Röhrchen NMR-Röhrchen
Complete with 2 PTFE-adapters to fir 5 cuvets or NMR tubes of 5 and 10 mm diameter in different lengths. Vacuum operated washers allowing the individual washing of 1 to 5 tube, complete with caps and FEPcappilaries (box 1 pce.).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge