Automatic burettes, Pellet pattern, class B
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3.114.506
WITG3.114.506EA 395 EUR InStock
WITG3.114.506
Automatic burettes, Pellet pattern, class B
Büretten
Automatic burettes, Pellet pattern, class B, made of amber borosilicate glass, short line graduation.
  • Stopcock, with hollow NS glass key, between burette and reservoir
  • Lateral stopcock, with solid NS glass key
  • Complete with 2000 ml reservoir bottle, made of DURAN® borosilicate glass, without aspirator bulb
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge