Bulb pipettes, one mark, class AS
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3.535.125 3.535.040 3.535.100.BL 3.535.200.BL 3.535.110 3.535.130 3.535.140 3.535.075 3.535.038 3.535.800 3.535.035 3.535.160 3.535.700 3.535.600 3.535.750 3.535.170 3.535.055
WITG3.535.125EA 91.8 EUR InStock
WITG3.535.125 WITG3.535.040 WITG3.535.100.BL WITG3.535.200.BL WITG3.535.110 WITG3.535.130 WITG3.535.140 WITG3.535.075 WITG3.535.038 WITG3.535.800 WITG3.535.035 WITG3.535.160 WITG3.535.700 WITG3.535.600 WITG3.535.750 WITG3.535.170 WITG3.535.055
Bulb pipettes, one mark, class AS
Pipetten Vollpipetten
These bulb pipettes are made of soda lime glass.
  • With amber or blue stain DIFFICO ring marks and inscriptions
  • With tempered tips
  • With ISO-COLOR-Code, acc. to DIN (ISO 648)
  • Waiting time 15 s
  • Calibrated to contain "EX"
  • Conformity certified
Zertifizierungen: DIN 12691
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge