Glass density bottles acc. Gay-Lussac
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
3.902.002
WITG3.902.002EA 28 EUR InStock
WITG3.902.002
Glass density bottles acc. Gay-Lussac
Pyknometer
Made of borosilicate glass.
Pycnometer with with ground-in capillary stopper, precisely adjusted to the second place of decimal, acc. to ISO 3507.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge