Dishes, evaporating, quartz glass
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
8.651.001 8.651.004 8.651.006 8.651.007 8.651.005
WITG8.651.001EA 0 EUR InStock
WITG8.651.001 WITG8.651.004 WITG8.651.006 WITG8.651.007 WITG8.651.005
Dishes, evaporating, quartz glass
Schalen/Schüsseln/Töpfe Abdampfschalen
Evaporating dishes, made of quartz glass, with spout.
  • Excellent chemical resistance
  • Excellent temperature resistance (up to 1000 °C)
Zertifizierungen: DIN 12341
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge