Low temperature BOD incubator, WiseCube®WIR
Lieferant: WITEG LABORTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
DH.WIR03250
WITGDH.WIR03250EA 4370 EUR InStock
WITGDH.WIR03250
Low temperature BOD incubator, WiseCube®WIR
Brutschränke BSB-Brutschränke
Low temperature BOD incubator with superior temperature uniformity by forced convection.
  • Intelligent control of refrigerator for preventing overload of compressor
  • RS232C interface for monitoring and controlling with PC
  • Stainless steel chamber and teflon-coated shelves, corrosion resistant
  • Over temperature and current protection, sensor error detection and leakage breaking
  • Back-light LCD display, alarm and timer available
  • Locking mode supported for experimental safety.
  • CFC-free (134A) refrigeration system
  • Digital fuzzy control system implementing superior temperature accuracy
  • Jog-shuttle control system
Used in animal and plant cell culture, constant temperature, germination test and BOD test.
Zertifizierungen: UL and CE certified.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge