Ion selective electrode system, VARiON® Plus 700 IQ
Lieferant: WTW ONLINE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
107040
WTWO107040EA 5490 EUR InStock
WTWO107040
Ion selective electrode system, VARiON® Plus 700 IQ
Elektroden ISE-Elektroden
In-situ combination sensor for direct measurements of ammonium and nitrate using only one sensor. Designed for the installation of a measuring electrode, a reference electrode and a compensation electrode.
  • Automatic compensation of interferences
  • Integrated NTC thermistor
  • Calibration-free, long time stable
  • Display shows the already compensated values, the potassium measuring value can be displayed optionally
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge