Confidence solution, 5580
Lieferant: YSI ENVIRONMENTAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
005580
YSIA005580EA 222 EUR InStock
YSIA005580
Confidence solution, 5580
Elektrolyte Elektrodenfülllösungen
This Confidence Solution® helps users to ensure probe performance of their pH, ORP, or conductivity probes.
This solution is designed for verifying measurements, not calibration. Simply immerse your multi-parameter probes in the solution and verify the pH, ORP, and conductivity values instantly. Ideal for quality checks during or after your study.
Expiration dates: Unopened=1 year, opened=3 months.
Lieferumfang: Case includes 6 plastic bottles (1 pint each).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge