Fast-response pH Sensor, 6589
Lieferant: YSI ENVIRONMENTAL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
606589
YSIA606589EA 944 EUR InStock
YSIA606589
Fast-response pH Sensor, 6589
Elektroden pH-Elektroden
Simple, accurate pH measurement in sensors designed for minimal maintenance in sampling, profiling, and long-term monitoring applications.
  • Faster response time in sampling and profiling applications
  • Maintains fast response time in Low Ionic Strength (LIS) and cold waters
  • Maintains fast response time throughout life of sensor
  • Stable readings and like-new performance in instruments with aging or contaminated connectors
YSI pH and pH/ORP sensors have been carefully designed to perform under all ionic strength conditions, from seawater with a conductivity of 53000 μS/cm, to 'average' freshwater lakes and rivers with conductivities of 200 to 1500 μS/cm, to pure mountain streams with conductivities as low as 15 μS/cm, which has historically been the most difficult medium with respect to accuracy, quick response to pH changes, and minimal flow dependence.
Use with: All 6-Series sondes with a user-replaceable pH port. Strengths: Provides rugged general monitoring flexibility. Produces  fast, stable readings in LIS Water and other difficult pH measuring environments. New design restores old instrument’s pH response time to like new.
Amplified sensor provides fast response and increased stability over standard pH sensors, despite the wear and tear of your instrument.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge