Digital camera, Axiocam MRc5
Lieferant: ZEISS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
0000000450354
ZEIS0000000450354EA 0 EUR InStock
ZEIS0000000450354
Digital camera, Axiocam MRc5
Kameras
This high resolution compact digital camera for microscopy can work in a stand-alone mode driver software, standard FireWire/IEEE1394 interface including cable and infrared barrier filter BG40.
  • Blue anodized aluminum, cooling fins, ¼" photo thread for tripod mount housing
  • One stage Peltier cooling control signals
  • 12 different capture modes and fast live images
  • Trigger in/out signal for control of external components
  • Fast live image modes for easy orientation on the specimen
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge