ZEN 3.9 Software for Microscopy Workstation
Lieferant: ZEISS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4101350002390
ZEIS4101350002390EA 0 EUR InStock
ZEIS4101350002390
ZEN 3.9 Software for Microscopy Workstation
Software Labor-Software
ZEN 3.9 software is an image acquisition and processor under Windows 10×64 user interface configurable.
  • Sheer control of the microscope systems and components
  • Extensive acquisition and analysis and results are provided as CZI image format
It includes with following functions such as arivis cloud advanced, basic 3D viewing, colocalization, connect basic, direct processing, manual EDF, measurement, panorama, spectral unmixing, ZEN data storage client and acquisition base (time series and multichannel).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge