Digital camera, Axiocam 305 mono
Lieferant: ZEISS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
4265609040000
ZEIS4265609040000EA 6920 EUR InStock
ZEIS4265609040000
Digital camera, Axiocam 305 mono
Kameras
Axiocam 305 mono is a CMOS microscope camera (C-mount) for routine fluorescence applications.
  • 5 MP sensor with 11 mm diagonal for good field of view
  • Fast readout with up to 36 full frame images for good live image experience and easy sample adjustment
  • Dark homogenous background helps to see even the finest structural details
  • Fast USB 3.0 connection, easy to install, with integrated camera power supply
  • Actively temperature stabilised sensor for optimised signal-to-noise and long term stability
  • Fast and efficient operation with ZEN imaging software
Recommended camera adapters: 0,5 × / 0,63 × / 1,0 ×
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge