Cleaning cloths, Praktitex® Pro
Lieferant: Zellstoff-Vertriebs-GmbH & Co. KG
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
10294-01
ZVGA10294-01EA 38.3 EUR InStock
ZVGA10294-01
Cleaning cloths, Praktitex® Pro
Wischtücher Reinigungstücher, wiederverwendbar Tücher für allgemeine Anwendungen
This microfibre cloth (80% polyester, 20% polyamide) has an open structure and is therefore ideal for cleaning heavily fouled surfaces.
  • Extremely fine fibres penetrate deep into the surface and thus effectively loosen adhering and grease-containing fouling
  • Washable and therefore reusable
Weight: 300 g/m²
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge