Hand-Operated Piston Dispensing Drum Pump, Polypropylene, 8 to 32 Strokes/gallon
Lieferant: Avantor Fluid Handling
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
06512-05
MFLX06512-05EA 39.1 EUR InStock
MFLX06512-05
Hand-Operated Piston Dispensing Drum Pump, Polypropylene, 8 to 32 Strokes/gallon
Pumpen Fass/Transferpumpen
VWR Item#MFLX06512-05
Your Organization's Price:
Economical piston pumps are ideal for a variety of fluids.
  • Adaptable to most pails and drums
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.
Other Products In this Family
  See all Items in this Product Family
TECHNISCHE DATEN
Key Features
  • Adaptable to most pails and drums
More About This Item
Models 06512-00 and -05 have adjustable strokes to allow for dispensing different amounts fo fluid with just one pump. Model 06512-00 pumps 64, 32, 22 and 16 strokes per gallon, model 60512-05 pumps 32, 16, 11 and 8 strokes per gallon.
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge