Gel permeation chromatography columns, MesoPore™
Lieferant: VARIAN
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PL1113-1325 PL1113-6325
VARIPL1113-1325EA 617 EUR InStock
VARIPL1113-1325 VARIPL1113-6325
Gel permeation chromatography columns, MesoPore™
Chromatographiesäulen
MesoPore columns have been specifically designed to give optimum results in the analysis of prepolymers, i.e. polymeric materials with a large oligomeric component.
  • Full solvent compatibility with no detrimental effect on efficiency
  • Low particle size extracts maximum information from the analyte
  • No MWD dislocations so the distribution is a faithful representation of the sample
By combining a 3 µm particle size with high pore volume, MesoPore columns give the highest resolution separations for the analysis of low molecular weight polymers, such as prepolymers, resins, polyols and siloxanes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge