Affinity chromatography columns, MBPTrap™ HP
Lieferant: Cytiva
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
28-9187-80 28-9187-79 28-9187-78 29-0486-41
28-9187-80EA 1210 EUR InStock
28-9187-80 28-9187-79 28-9187-78 29-0486-41
Affinity chromatography columns, MBPTrap™ HP
Chromatographiesäulen
MBPTrap™ HP columns prepacked with Dextrin Sepharose™ High Performance provide convenient, high binding capacity, affinity purification of recombinant proteins tagged with maltose binding protein (MBP).
  • Highly pure MBP-tagged recombinant proteins eluted in concentrated form and small volumes
  • Compatible with commonly used buffers and easily regenerated with 0,5 M NaOH
  • Physiological conditions and mild elution preserve the activity of the target protein
  • Convenient, time-saving operation and reproducible results
Dextrin Sepharose High Performance has high binding capacity for MBP-tagged proteins. This robust and stable Sepharose High Performance based resin enables elution of target protein in narrow peaks, which minimizes the need for further concentration steps.
Purification is performed under physiological conditions and the mild elution preserves the activity of the target protein. The mild conditions even allow purification of intact protein complexes. Due to the high specificity of the binding, very high purity of the eluted protein is achieved in just one step. Dextrin Sepharose High Performance and MBPTrap HP are easily regenerated using 0,5 M NaOH.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge