LONZ17-942
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
LONZ17-942EEA 0 EUR InStock
LONZ17-942E
LONZ17-942
Trypan Blue Stain is a stain used to distinguish viable from nonviable cells. Nonviable cells will absorb the dye Trypan Blue and appear blue, while viable cells will exclude the dye.
Cells should be in a single cell suspension in buffered saline before counting.
Since Trypan Blue has a higher affinity for serum protein than for cellular proteins, suspending the cells in medium containing serum will generate a dark background.
Commonly used to assess cell viability.
Order Now
PRODUKT
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge