Disinfectant, non sterile chlorinated alkaline sanitiser, Amber
Lieferant: TRISTEL SOLUTIONS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
CRY502 CRY503
TRSTCRY502EA 0 EUR InStock
TRSTCRY502 TRSTCRY503
Disinfectant, non sterile chlorinated alkaline sanitiser, Amber
Reinigungsmittel Desinfektionsmittel
A non foaming, chlorine based, alkaline sanitiser.
  • Ideal for use on walls, floors, equipment and product contact areas
  • Suitable for manual use
  • Dilution for use: 0,5 to 5% v/v
Areas of use include walls, floors, product contact areas and equipment such as filling lines. Amber is not soft metal safe, but can be used on a variety of surfaces including stainless steel and is also ideal for drain and mop disinfection.
All disinfectants are supported by comprehensive technical files which provide all the documentation for validation. All disinfectants are supported by the BPD and have independent efficacy data to prove performance.
Zertifizierungen: EN standards: EN1276 - bactericidal activity, EN1650.
Lieferumfang: Supplied with its own certificate of analysis. A detailed technical file (including: product specification, manufacturing process, shelf life, microbiological and analytical data) is available on request.
Verpackung: Available in 20, 200 and 1000 litre pack sizes.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge