Superficially porous particle (SPP) columns, Brownlee™ C8
Lieferant: PERKIN ELMER GC INSTRUMENTS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
N9308422 N9308434
PERCN9308422EA 614 EUR InStock
PERCN9308422 PERCN9308434
Superficially porous particle (SPP) columns, Brownlee™ C8
Chromatographiesäulen
These superficially porous particle (SPP) columns produce sharper peaks and faster separation results due to their breakthrough particle design and size.
  • General purpose octyl phase for reversed phase separations with less retension than C18
  • Exhibits peak shape for wide range of compounds
  • pH range of 2 to 9 and the temperature range of 60 °C
The design allows for a shorter diffusion path, reducing the time solute molecules spend inside the particles while passing through the stationary phase.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge