Hand cleanser dispensers, Plus
Lieferant: B. Braun
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
1418894 1418895 1418892 1418893
BRAU1418894EA 180 EUR InStock
BRAU1418894 BRAU1418895 BRAU1418892 BRAU1418893
Hand cleanser dispensers, Plus
Seifen- und Flüssigseifenspender
Wall mounted hand cleanser dispensers for contact free hygiene applications.
  • Control lever of two different lengths
  • Easy and fast pump cleaning by front removal
  • Flexible seal cap to avoid evaporation and two interchangeable clips - disinfectant/cleanser
Suitable for use with 'standard' 500 and 1000 ml containers.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge