Shorts, Crafts 201 FAS
Lieferant: FRISTADS KANSAS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
100276940C44 100276940C46 100276940C48 100276940C50 100276940C52 100276940C54 100276940C56 100276940C58 100276940C60 100276940C62 100276941C44 100276941C46 100276941C48 100276941C50 100276941C52 100276941C54 100276941C56 100276941C58 100276941C60 100276941C62
KWIN100276940C44EA 77.9 EUR InStock
KWIN100276940C44 KWIN100276940C46 KWIN100276940C48 KWIN100276940C50 KWIN100276940C52 KWIN100276940C54 KWIN100276940C56 KWIN100276940C58 KWIN100276940C60 KWIN100276940C62 KWIN100276941C44 KWIN100276941C46 KWIN100276941C48 KWIN100276941C50 KWIN100276941C52 KWIN100276941C54 KWIN100276941C56 KWIN100276941C58 KWIN100276941C60 KWIN100276941C62
Shorts, Crafts 201 FAS
Arbeitskleidung
Durable short trousers made of FAS®, Fristads best twill consisting of 100% cotton. Ideal for people working in building and construction.
  • Rounded knees, Cordura® reinforced knee pockets with opening for height-adjustable knee pads
  • Two loose-hanging, tuckable front pockets, one with extra pocket, one with three small pockets and tool loops
  • Two front pockets, two large back pockets, folding ruler pocket over side seam button and loop for sheath knife and pen pocket
  • Mobile phone pocket with flap, leg pocket with flap and concealed ID-card pocket
  • Two hammer loops, D-ring
Weight: 375 g/m²
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge