Filter für X-plore® Rd90
Lieferant: DRAEGER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
6736705 6738870 6738865
DRAE6736705EA 43 EUR InStock
DRAE6736705 DRAE6738870 DRAE6738865
Filter für X-plore® Rd90
Atemschutzmasken Mehrweg-Atemschutzmasken Maskenfilter
The filters of the Dräger X-plore® Rd90 series protect from hazardous gases and vapors, have a long life and can be used for many different masks.
  • Filters have been designed for use in the Dräger half masks, X-plore® 4390 and 4790. With the 40/90 filter box adapter they can also be used for any mask with a Rd40 connection in accordance with EN 148-1.
  • Gas filters and combination filters have a shelf-life of 6 years and, the particle filters a shelf life of 12 years from date of manufacture
  • Filters are packed individually to ensure optimum protection of the unused filter
  • Aluminium housing ensures that any damage to the filter is easily detected, therefore providing extra security
The filters are suitable for use with half and full face masks with standard thread connection Rd40 according to EN 148-1.
Zertifizierungen: EN 14387, EN 143, EN 148-1
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge