Storage cabinets for acids and alkalis, SLS 1200
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
B80-3016-D B80-3023-D B80-3017-D
LACOB80-3016-DEA 0 EUR InStock
LACOB80-3016-D LACOB80-3023-D LACOB80-3017-D
Storage cabinets for acids and alkalis, SLS 1200
Möbel Lagerschränke
These storage cabinet body made from HPL-coated sheets, particularly impact, scratch and abrasion proof are used for storage of acids and alkalis.
  • Doors with separate cylinder locks
  • Levelling feet for installing cabinet horizontally
  • Air supply by ventilation grille in the back wall
  • Colour RAL 7035 - light grey
  • Ventilation connection for exhaust air extraction on cabinet roof
Inner body and inner fittings completely made from corrosion-proof components.
It features with central horizontal division for storage of acids and alkalis in four separate compartments.
Cabinet with inner fittings has pull-out shelves with end stop, front edge of shelves in attractive solid wood design and PE sump tray on shelf secured against slipping and easy to remove.
Zertifizierungen: Conforms with TRGS 526 / DGUV, ASR A1,3.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge