Reversed phase HPLC columns, ProSwift™ RP
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
069477 064296 064297 064298 066640
555-7121EA 1010 EUR InStock
555-7121 555-7143 555-7144 555-7145 555-7157
Reversed phase HPLC columns, ProSwift™ RP
Chromatographiesäulen
Reversed phase columns provide the advantages of high resolution at high flow rates for fast protein separation and analysis.
  • High resolution at high speed
  • Operational flow rates available
  • Optimal performance in a board range of applications
  • pH range 1 to 4
  • High loading capacity
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge