HPLC columns, Acclaim™ Phenyl-1
Lieferant: Thermo Fisher Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
071969 071970 071971 071973 071974 079697
555-7171EA 1010 EUR InStock
555-7171 555-7172 555-7173 555-7175 555-7176 555-7528
HPLC columns, Acclaim™ Phenyl-1
Chromatographiesäulen
These columns are widely used in pharmaceutical, environmental, food testing and product quality testing and it is suitable for the analysis of the aromatic analytes.
  • High aromatic selectivity
  • High hydrophobic retention
  • Compatibility with high aqueous mobile phase
  • High efficiency and rugged packing
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge