CombiPAL headspace syringes
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
G6500-80109 G6500-80107 G6500-80111
HEWLG6500-80109EA 0 EUR InStock
HEWLG6500-80109 HEWLG6500-80107 HEWLG6500-80111
CombiPAL headspace syringes
Spritzen Spritzen für die Chromatographie
Fixed needle headspace syringes feature a side port needle for gas flushing.
Micro seal is used to minimise instrument downtime and to prevent lost or compromised data caused by septum leaks and liner contamination.
Zertifizierungen: Conforms with strict CTC standards.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge