Pipette controller, Pipet-Aid® XP2
Lieferant: DRUMMOND
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
4-000-501-I 4-000-501-UK
DRUM4-000-501-IEA 1430 EUR InStock
DRUM4-000-501-I DRUM4-000-501-UK
Pipette controller, Pipet-Aid® XP2
Pipetten Pipettierhelfer
This controller is an advanced version of the Pipet-Aid® XP controller and accepts 1 to 100 ml pipettes.
  • New ergonomic design
  • New power source for uninterrupted extended operation; can be charged while in use
  • Ultra quiet precision pump provides great control for aspiration or dispensing
Lieferumfang: The unit is supplied complete with a power supply/charger, four extra filters, and a removable stand.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge