Storage cabinets for drums, WSP
Lieferant: LACONT UMWELTTECHNIK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
P21-2482-A P21-2480-A P21-2481-A P21-2483-A
LACOP21-2482-AEA 0 EUR InStock
LACOP21-2482-A LACOP21-2480-A LACOP21-2481-A LACOP21-2483-A
Storage cabinets for drums, WSP
Möbel Lagerschränke
Storage cabinets are used for the storage of water-endangering liquids.
  • Base sump tray made from 3 mm sheet steel
  • Ground clearance 100 mm for internal transport
  • Corrosion protection by primer coating and painting of sump pallet
  • Compartment for documents in the doors as standard
Weather-proof superstructure made from galvanised sheet steel, with venting slots in the side walls.
Zertifizierungen: Conforms with WHG.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge