Hotplates, OTS 40
Lieferant: MEDITE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
01-4002-00 01-4003-00 01-4001-00 01-4004-00 01-4002-01
632-7001EA 1130 EUR InStock
632-7001 632-0050 632-0059 632-0025 MEDT01-4002-01
Hotplates, OTS 40
Heizgeräte Heizplatten
OTS 40 is a black plastic coated aluminium plate that serves to achieve the desired temperature and keep it constant.
  • Adjustable
  • Controllable
  • Digital display indication
Durability and high stability are also important factors for research labs and chemical industry to choose these tables, which provides both at a high-quality standard.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge