Sicherheitsschuhe, zum Schnüren, Bacou Original Siluo
Lieferant: Honeywell Safety Products
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Bacou Original Siluo
DALL6246109/39EA 82.9 EUR InStock
DALL6246109/39 DALL6246109/48 DALL6246109/46 DALL6246109/47 DALL6246109/44 DALL6246109/45 DALL6246109/42 DALL6246109/43 DALL6246109/40 DALL6246109/41
Sicherheitsschuhe, zum Schnüren, Bacou Original Siluo
Schuhe und Stiefel
Safety shoes made from suede leather and ventilated mesh upper, antibacterial 3-D lining plus a PU sole.
  • Dual-density PU outsole guarantees stability and shock absorbency
  • Stainless steel, puncture resistant midsole covers most extensive panel of risk
  • Light, metal-free Spacium™ toecap in injected polymer (200 J)
  • Extra wide toecap for great comfort, suits all foot types
  • Lateral bellows prevents dust from entering the shoe
Colour: Grey/orange
Zertifizierungen: EN ISO 20345:2011; S1P, HI, CI, SRC.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge