Test tubes, round bottom
Lieferant: DELTALAB
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
916150 818150 820150 801075 801275 901275 913100 816150 916100
212-3308EA 145 EUR InStock
212-3308 212-3310 212-3312 212-3315 212-3317 DELT901275 DELT913100 DELT816150 DELT916100
Test tubes, round bottom
Röhrchen Reagenzgläser- und Probenröhrchen
Tubes are made of borosilicate or soda glass.
  • Good chemical resistance
  • Good temperature resistance
High quality of tubes is reflected in uniformity of wall thickness, diameter and height dimensions.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge