TZe Labelling tape cartridges for P-Touch
Lieferant: BROTHER
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
TZE-441 TZE-325 TZE-721 TZE-222 TZE-211 TZE-551 TZE-252 TZE-354 TZE-231 TZE-111 TZE-315 Z243 TZE-251 TZE-242 TZE-241 TZE-335 TZE-232 TZE-345 TZE-223 TZE-221 TZE-151 TZE-141 TZE-132 TZE-131 TZE-121 TZE-233 TZE-621 TZE-C31 TZE-B31 TZE-741 TZE-731 TZE-651 TZE-631 TZE-541 TZE-531 TZE-435 TZE-431 TZE-421 TZE-641 TZE611 TZE-135 TZE-S231 TZE-MQP35
811-0888EA 913 EUR InStock
811-0888 811-0879 811-0901 811-0866 811-0864 811-0894 811-0875 811-0884 BROTTZE-231 BROTTZE-111 BROTTZE-315 BROTZ243 BROTTZE-251 BROTTZE-242 BROTTZE-241 BROTTZE-335 BROTTZE-232 BROTTZE-345 BROTTZE-223 BROTTZE-221 BROTTZE-151 BROTTZE-141 BROTTZE-132 BROTTZE-131 BROTTZE-121 BROTTZE-233 BROTTZE-621 BROTTZE-C31 BROTTZE-B31 BROTTZE-741 BROTTZE-731 BROTTZE-651 BROTTZE-631 BROTTZE-541 BROTTZE-531 BROTTZE-435 BROTTZE-431 BROTTZE-421 BROTTZE-641 BROTTZE611 BROTTZE-135 BROTTZE-S231 BROTTZE-MQP35
TZe Labelling tape cartridges for P-Touch
Etiketten
These TZe labelling tapes are developed for use with P-Touch labelprinters. They feature the exclusive laminated tape process and standard adhesive. Best suited for smooth flat surfaces.
  • Perfect for everyday applications
  • Stay on under normal indoor and outdoor use, including hot and cold environments
Length: 8,0 m
Different colours available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge