Surface disinfectant, perform® sterile concentrate QB
Lieferant: Schülke
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SCUE112703EA 235 EUR InStock
SCUE112703
Surface disinfectant, perform® sterile concentrate QB
Reinigungsmittel Desinfektionsmittel
Surface disinfectant for cleanrooms provided as a concentrated liquid, based on quaternary ammonium compounds and biguanide.
  • Fast and effective
  • Gamma irradiated and microbial filtered (0,2 µm)
  • Convenient single-use bottle
  • Colour- and perfume-free
  • Bactericidal, virucidal (BVDV, Vaccinia), yeasticidal
Achtung! Bitte beachten: Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge