Columns, ACQUITY UPLC™ CSH C18
Lieferant: WATERS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
186005297 186005298 186005296
WATE186005297EA 1970 EUR InStock
WATE186005297 WATE186005298 WATE186005296
Columns, ACQUITY UPLC™ CSH C18
Chromatographiesäulen
ACQUITY UPLC method validation kit includes three batches of chromatographic media (derived from different base particles) to judge the quality, reliability, and consistency of the chromatographic method.
  • Exceptional batch-to-batch reproducibility
  • Broad range in selectivity
  • Excellent stability at low pH
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge