Combined pH electrodes, 405-DPAS
Lieferant: Mettler - Toledo
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
59903230 59903233 59903231 59903232 59903234
METN59903230EA 0 EUR InStock
METN59903230 METN59903233 METN59903231 METN59903232 METN59903234
Combined pH electrodes, 405-DPAS
Elektroden pH-Elektroden
405-DPAS is a pre-pressurized, gel-filled, low maintenance pH probe, specially developped for in-line measurements in biotech and food and beverage applications.
  • Filled with gel-reference electrolyte which is pre-pressurized to 2,5 bar (36 psi)
  • Internal over-pressure ensures gradual electrolyte outflow efficiently cleaning the diaphragm and preventing influx of process medium
  • Probe is further provided with a patented silver-ion trap to prevent diaphragm fouling in sulfide-containing media and prolonging the operative life of the probe
Designed for use in reactors, tanks, open basins and transfer lines under harsh chemical process conditions.
Reference system: Argenthal system
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge