Examination gloves, nitrile, Meditrade® Nitril 3000
Lieferant: MEDITRADE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
1280L 1280M 1280S 1280XS 1280XL
ROES1280LEA 17.1 EUR InStock
ROES1280L ROES1280M ROES1280S ROES1280XS ROES1280XL
Examination gloves, nitrile, Meditrade® Nitril 3000
Handschuhe
Disposable examination gloves for environments with high contamination risks. The gloves are resistant against a range of chemical solvents.
  • Non sterile, smooth finish
  • Ambidextrous
  • Skin-friendly, latex-, protein- and powder-free
Colour: White
EN 455; AQL 1,5
Zertifizierungen: EN 455; AQL 1,5
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge