Jerricans, narrow neck, HDPE
Lieferant: LAMAPLAST
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
3952 3951 3950 3949 3948
LAMA3952EA 38.2 EUR InStock
LAMA3952 LAMA3951 LAMA3950 LAMA3949 LAMA3948
Jerricans, narrow neck, HDPE
Flaschen Kanister
HDPE, translucent
  • Supplied with pouring spout (except for 5 L jerrican)
  • Spout is long enough for accurate pouring of liquids
  • With tethered sealing cap that cannot be lost
Universal canister for industry, trade and laboratories. Ideal for packaging, storage and transportation.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge