SUPELCO Vial Racks
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
23205-U 23207 23201 23202 23204-U 23206
SIAL23205-UEA 161 EUR InStock
SIAL23205-U SIAL23207 SIAL23201 SIAL23202 SIAL23204-U SIAL23206
SUPELCO Vial Racks
Gestelle Gestelle für Chromatographie-Ampullen
PP, red.
  • Can be stacked when empty or full
  • Autoclavable 120 °C
  • Very good chemical resistance
Economical and convenient storage for samples in screw cap and crimp seal vials.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge