Gas cylinder supplies
Lieferant: Agilent
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
5183-1941 5183-4641 5183-4642 5183-4643 5183-4644 5183-4645
HEWL5183-1941EA 296 EUR InStock
HEWL5183-1941 HEWL5183-4641 HEWL5183-4642 HEWL5183-4643 HEWL5183-4644 HEWL5183-4645
Gas cylinder supplies
Chromatographie-Zubehör Chromatographie-Ersatzteile
A broad spectrum of economical brass regulators, wall brackets, and NPT adapters.
  • Maximum inlet pressure: 206,8 bar
  • Operating temperature range: −40 to 165 °F
Dual-stage regulators provide more precise and consistent pressure control than single-stage regulators.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge