Conductivity meters, bench, SensION™+ EC7 / EC71
Lieferant: Hach
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
LPV3070980002 LPV3170980002
HACHLPV3070980002EA 1184 EUR InStock
HACHLPV3070980002 HACHLPV3170980002
Conductivity meters, bench, SensION™+ EC7 / EC71
Elektrochemische Messgeräte Leitfähigkeitsmessgeräte
These all-in-one systems with guided menu navigation make general water quality meter testing fast and simple.
  • Simple and fast measurements
  • Measure electrical conductivity and salinity (NaCl), the GLP meter (EC71) also measures total dissolved solids (TDS)
  • Data Logger for 400 measurements, calibration data and cell record (Only GLP meter EC71)
  • Data can be sent to printer or PC through the RS 232 C interface (Only GLP meter EC71)
Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing. A guided navigation menu allows you to follow simple prompts to set up your measurements thus reducing the time necessary to start testing, and improving the accuracy of measurements.
Lieferumfang: The sensION+ EC7 benchtop conductivity kit is supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, sensION+ 5070 conductivity cell with temperature sensor, 147 µS/cm, 1413 µS/cm, 12,88 mS/cm conductivity standard solutions, 3 calibration beakers and printed documentation.
The sensION+ EC71 (GLP) meter is supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, 147 µS/cm, 1413 µS/cm, 12,88 mS/cm conductivity standard solutions, 3 calibration beakers and printed documentation.
The EC71 kit additionally includes the sensION+ 5070 conductivity cell with temperature sensor.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge