Anti-IgG Donkey Polyclonal Antibody (CF™ 770)
Lieferant: Biotium
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
20279 20279-1
20279.EA 359 EUR InStock
20279. 20279-1
Anti-IgG Donkey Polyclonal Antibody (CF™ 770)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF® 770 Dye
Epitope:
Host: Donkey
Isotype: none
Reactivity: Human
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge