School kit for environmental microbiology
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SART24002EA 1170 EUR InStock
SART24002
School kit for environmental microbiology
The school kit for microbiological experiments is an ideal teaching aid for instruction in microbiology and environmental protection in schools and other educational institutes. The rugged aluminum case contains all the equipment necessary for microbiological testing. The handbook included in the case provides general instructions and detailed descriptions of methods for seven experiments: Detection of microorganisms in water, air, and soil; the effects of antibiotics; detection of yeasts on substrates in nature; production of gas through alcoholic fermentation; and bacterial growth at different temperatures.The vacuum, which is necessary for the filtration, is created with help of a syringe and a three way valve.
  • Seven experiments - easy to understand
  • Compact and robust design
  • Portable
Lieferumfang: Kit includes two filtration systems, tweezers, culture media; inoculating loop, filter pads; instructions for seven experiments.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge